Laboratory Cold Rooms, Precision Controlled Environments, Darwin Chambers Company
Supplier: Darwin Chambers Company
Certificates
About this item
Darwin Laboratory Cold Rooms feature reliable, efficient environmental controls for a variety of applications
- All rooms are constructed using 10.2 cm (4") insulated panels, R Value 31+, with white embossed aluminum interior surface finishes and white embossed galvanized steel exterior finishes
- All include a 5.08 cm (2") insulated smooth aluminum or stainless steel floor, capable of accommodating up to 317.5 kg (700 lbs.) of evenly distributed weight
- These rooms are modular, featuring "tongue-in-groove" construction
- All major components are pre-wired, pre-brazed, and pre-assembled, ready for quick onsite assembly; Darwin PFTCU liquid-circuited cooling
Suitable uses include GMP storage and cold storage for seed and agricultural products, nutritional supplements, and more.
Temperature Control at Sensor: ±0.2 °C, sensor (100 ohm Platinum RTD) accuracy: ±0.2 °C
Certifications: Panels are UL listed, and conform to nationally recognized industry standards.
Ordering information: Installation services are not included; please call for quote. Optional accessories are available separately.
Frequently Bought Together