Rodent Nesting Sheets™, Certified, Bio-Serv
Supplier: Bio-Serv
Certificates
About this item
Rodent Nesting Sheets are pure, made of 100% virgin wood pulp
Dispenser is made of 100% sturdy acrylic. Clear with removable lid. Holds one sleeve (1,400) of nesting sheets. Dispenses one sheet at a time. Mounts to wall or free-standing option. Hand wash with soap and water only.
The lint-free sheets do not cause eye irritation and are a safer alternative to cotton nesting material. Encourages natural nest building behavior. Will not clog automated disposal systems. Autoclave like any other paper or cotton products.
Certifications: Certified (Contaminant Screened).
Frequently Bought Together