Fruit and Nut Foraging Mix, Certified, Bio-Serv
Catalog # 89067-313
Supplier: Bio-Serv
Specifications
- Description:Fruit and Nut Foraging Mix
- Environmentally Preferable:
- Cat. No.:89067-313
- Packaging:5 kg/Box
- Supplier no.:F3084-5
Specifications
About this item
Delicious, natural foraging mixture.
- Use with Tuff Turf Foraging Board or scatter into bedding
- Contains no shells
- Nutritionally assayed
- Shelf Life: 6 months, cool dry storage
Ingredients: Peanuts, Sunflower Seeds, Banana Chips and Raisins.
Caloric value: 5.26 kcal/g
Nutritional ash content: 2.6%
Nutritional carbohydrate content: 32.4%
Nutritional fat content: 36.0%
Nutritional fibre content: 3.0%
Nutritional moisture content:<10%
Nutritional protein content: 18.0%
Certifications: Certified (Contaminant Screened).