Order Entry
United States
ContactUsLinkComponent
VWR® 4-Place Mini Bead Mill Homogenizer
VWR® 4-Place Mini Bead Mill Homogenizer
Catalog # 10158-558
Supplier:  VWR International
undefined
VWR® 4-Place Mini Bead Mill Homogenizer
Catalog # 10158-558
Supplier:  VWR International

Specifications

  • Description:
    VWR® 4-Place Mini Bead Mill
  • Environmentally Preferable:
  • Processing Range:
    25 µL to 5 mL
  • Electrical:
    100–230V, 50/60 Hz
  • Run Time:
    1 sec. to 5 min.
  • Dimensions:
    21.5 x 29.2 x 25.4 cm (8.5 x 11.5 x 10")
  • Weight:
    7.48 kg (16.5 lbs.)
  • Cat. No.:
    10158-558

Specifications

About this item

The VWR® 4-Place Mini Bead Mill Homogenizer is a compact and powerful bead mill homogenizer.

  • Specifically designed for grinding, lysing, and the homogenization of biological samples prior to molecular extraction
  • Ideal solution for releasing DNA, RNA, proteins, and enzymes from tough samples while still retaining molecular integrity
  • Offers a broad performance range

Using sample tubes pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization in less than 60 seconds. The VWR® 4-Place Mini Bead Mill supports simultaneous processing of samples in 4x0.5 ml, 4x1.5 ml, 4x2 ml, or 1x7 ml tubes. It does not require a cool down period between runs, allowing for non-stop use and high throughput. The unit features a convenient front loading design and safety lid closure.

Certifications: CE marked.
Ordering information: Includes the VWR® Mini Bead Mill Drive Unit, integrated 0.5 ml, 1.5 ml and 2 ml tube carriage, interchangeable 1x7 ml tube carriage, user manual, and two-year warranty. VWR® disposable bead tubes are sold separately.