To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Valosin Containing Protein
- Antigen Symbol:VCP
- Clonality:Monoclonal
- Clone:2A4B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:ALS14|HEL-S-70|IBMPFD1|TERA|p97|HEL-220|IBMPFD
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:VCP
- Purification:Protein G purification
- Cat. No.:10155-902
- Supplier no.:60316-1-IG
Specifications
About this item
The VCP antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human VCP. This antibody recognizes human, mouse antigen. The VCP antibody has been validated for the following applications: ELISA, WB, IHC analysis.
WB 1 : 1000
Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: 2A4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse