To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ATP/GTP binding Protein-like 3
- Antigen Symbol:AGBL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:AGBL3
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:AGBL3
- Purification:Antigen affinity purification
- Cat. No.:10154-714
- Supplier no.:16990-1-AP
Specifications
About this item
The AGBL3 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human AGBL3. This Antibody recognizes human,mouse,rat Antigen. The AGBL3 Antibody has been validated for the following applications: ELISA, WB, IHC analysis.
IHC 1 : 1200
Type: Primary
Antigen: AGBL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat