Order Entry
United States
ContactUsLinkComponent
RNA Gel Buffer 10X (MOPS buffer), Quality Biological
RNA Gel Buffer 10X (MOPS buffer), Quality Biological
  10128-502
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
RNA Gel Buffer 10X (MOPS buffer), Quality Biological
  10128-502
 :  351-059-101
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  

 

  • Description:
    RNA Gel buffer 10X
  • Size:
    1× 500 ml
  • Environmentally Preferable:
  • Sterility:
    Sterile filtered
  • Test Description:
    DNase, RNase tested
  • Shelf Life:
    36 months
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    10128-502
  • pH:
    7.0± 0.1
  • Supplier no.:
    351-059-101

 

 

MOPS buffer is used at 1X for the electrophoresis of RNA as the running buffer to separate RNA samples on agarose and formaldehyde-agarose (denaturing) gels. MOPS buffer is also used for Northern Blots -the hybridization of total RNA or mRNA to a membrane.

  • Liquid, 10X concentration
  • Filtered through a 0.22 µm membrane
  • DNase, RNase tested

MOPS buffer is suitable for use in RNA Gel Electrophoresis, Northern Blot Analysis, and when making formaldehyde-agarose gels and associated running buffer for RNA electrophoresis.

 : Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
 : Research use only.