Sodium dodecyl sulfate (SDS) 10% in aqueous solution, sterile filtered
Supplier: QUALITY BIOLOGICAL, INC.
Certificates
About this item
Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Density:1…1.1 g/cm³ (20 °C)
Molecular Weight:288.38 g/mol
Merck Index:14,08636
Storage Temperature:Ambient
EINECS:205-788-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together