About this item
For shaking in ambient conditions: on the bench, in an incubator or in a warm or cold room, the Innova Platform Shakers are the instrument of choice
Quiet operation provides a favorable work enviornment. Shaking speeds between 25 – 500 rpm (±1 rpm) provides versatility for culturing a wide range of cell types. Includes audible and visual alarms, timer (0 – 99.9 h) with continuous operation or automatic shut-off at the end of the study.
Dependable operation is due in large part to the New Brunswick triple-eccentric counterbalanced drive. The Innova platform shaker model 2050 comes with additional stabilizing feet and counterweight to balance larger loads on an oversized platform. Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years.