About this item
Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors
- Total Protein= 4.0 - 7.0 gm/dL
- pH: 6.5 - 8.0
- Presentation: Frozen Brown Solution
In immunohistochemistry, antibodies are used to detect the presence and abundance of certain antigens in a given tissue sample. The use of a blocking serum such as rabbit serum is important because it prevents nonspecific binding and interaction between the antibodies and non-targeted antigens. Rabbit Serum is also a common component in media used to maintain cell culture.
Rabbit Serum Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry
Total Protein= 4.0 - 7.0 gm/dL
Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- IC19135780
- Storage Temperature:
- -20 °C