To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Acidic Nuclear Phosphoprotein 32 Family, Member B
- Antigen Symbol:ANP32B
- Clonality:Monoclonal
- Clone:5A1F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:APRIL|PHAPI2|SSP29
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. No.:10082-948
- Supplier no.:66160-1-IG
Specifications
About this item
ANP32B is a member of the highly conserved acidic leucine-rich nuclear phosphoprotein 32 (ANP32) family. The ANP32 proteins have been implicated in a broad array of physiological processes, including cell differentiation, apoptotic cell death, and cell proliferation. ANP32B was identified as a novel direct substrate for caspase-3 and acted as a negative regulator for leukemic cell apoptosis. ANP32B controls the expression of the dendritic cell maturation factor CD83 by regulating the transport of its mRNA to the cytoplasm. It also modulates the activity of the transcription factor Kruppel-like factor 5 (KLF5).
Western Blot: HEK-293 Cells, 1:500-1:5000
Type: Primary
Antigen: ANP32B
Clonality: Monoclonal
Clone: 5A1F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human