Order Entry
United States
ContactUsLinkComponent
 
Anti-PAX1 Mouse Monoclonal Antibody [clone: 5A10D9]
  10090-934
undefined
Anti-PAX1 Mouse Monoclonal Antibody [clone: 5A10D9]
  10090-934
 :  60217-1-IG-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Paired Box 1
  • Antigen Symbol:
    PAX1
  • Clonality:
    Monoclonal
  • Clone:
    5A10D9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Protein A Purification
  • Cat. No.:
    10090-934
  • Supplier no.:
    60217-1-IG

 

 

PAX1, also named as HUP48, is a transcriptional activator. It may play a role in the formation of segmented structures of the embryo and may play an important role in the normal development of the vertebral column. It is a marker of the sclerotomal compartment. PAX1 was reported that, quantitative measurement of PAX1 hyper-methylation in cervical scrapings is highly sensitive and is more specific than HC2 in detection of cervical cancer. It is expressed by vSMC and pericytes during the earliest phases of vascular wall formation. This antibody is specific to PAX1.

Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: PAX1
Clonality: Monoclonal
Clone: 5A10D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse