To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Splicing factor, arginine/serine-rich 8
- Antigen Symbol:SFRS8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:Splicing factor|arginine/serine-rich 8|SWAP|SFSWAP|SFRS8|Suppressor of white apricot protein homolog|suppressor of white-apricot homolog
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10094-464
- Supplier no.:24705-1-AP
Specifications
About this item
SFRS8, also named as SFSWAP and SWAP, is splicing regulator. It regulates the splicing of fibronectin and CD45 genes. SFRS8 represses the splicing of MAPT/Tau exon 10. This antibody detects 130-150 kDa SFRS8 protein.
Western Blot:Jurkat Cells, 1:1000-1:10000
Type: Primary
Antigen: SFRS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human