To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Alport syndrome, mental retardation, midface hypoplasia And elliptocytosis chromosomal region gene 1
- Antigen Symbol:AMMECR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:AMMECR1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-840
- Supplier no.:24687-1-AP
Specifications
About this item
AMMECR1 also named as AMME syndrome candidate gene 1 protein is a 333 amino acid protein, which contains 1 AMMECER1 domain. Containing a glycine-rich N terminus, the AMMECR1 protein exhibits putative nuclear localization and a substantial level of instability, suggesting it plays a role in regulation. The deletion of AMMECR1 gene may be involved in glomerulonephritis, sensorineural hearing loss, mental retardation, midface hypoplasia and elliptocytosis.
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human cerebellum Tissue, 1:20-1:200
Type: Primary
Antigen: AMMECR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse