Order Entry
United States
ContactUsLinkComponent
Anti-SPRED2 Rabbit Polyclonal Antibody
Anti-SPRED2 Rabbit Polyclonal Antibody
Catalog # 10095-122
Supplier:  Proteintech
undefined
Anti-SPRED2 Rabbit Polyclonal Antibody
Catalog # 10095-122
Supplier:  Proteintech
Supplier Number:  24091-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Sprouty-Related, EVH1 Domain Containing 2
  • Antigen Symbol:
    SPRED2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    EVH1 domain-containing protein 2|Spred-2|Sprouty-related
  • Storage Buffer:
    PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen Affinity Purified
  • Cat. No.:
    10095-122
  • Supplier no.:
    24091-1-AP

Specifications

About this item

SPRED2 is a member of the SPRED family of proteins that modulate growth factor receptor signaling by inhibiting the Ras-MAPK cascade . SPRED family members are characterized by an N-terminal Enabled/VASP homology 1 domain (EVH1), a central c-Kit binding domain (KBD) and a C-terminal Sprouty-related domain (SPR) . Three mammalian SPREDs (SPRED1-3) have been described. In adult tissues, SPRED2 is expressed ubiquitously, but most highly expressed in glandular epithelia . Reduced expression level of SPRED2 has been reported in human hepatocellular carcinoma (HCC), suggesting SPRED2 may play a role in tumorgenesis . Gene disruption of SPRED2 causes dwarfism .

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: SPRED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat