To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:PR Domain zinc Finger protein 1
- Antigen Symbol:PRDM10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:PFM7
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10092-754
- Supplier no.:23827-1-AP
Specifications
About this item
PRDM10 is a member of PRDM family, which contains a PR(PRDI-BF1 and RIZ homology) domain, and PRDM is also a subgroup of PR/SET transcription factor family for its PR domain is similar to the catalytic motif of the SET domain of histone methyltransferase(HMTase). Thus PRDM is suggested involve in the regulation of genome expression and chromatin remodeling. PRDM10 shares characteristics with the PR/SET faimly and clusters of zinc fingers DNA binding motifs. It's postualated PRDM10 has an essential role in regulating gene expression and tissue differentiation and a gene repressor. It may play a role in the pathogenesis of gangliosidosis.
Western Blot: HEK-293 Cells, 1:500-1:5000; IP: Mouse Testis Tissue, 1:200-1:2000
Type: Primary
Antigen: PRDM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse