Order Entry
United States
ContactUsLinkComponent
Anti-RPRD1A Rabbit Polyclonal Antibody
Anti-RPRD1A Rabbit Polyclonal Antibody
Catalog # 10090-930
Supplier:  Proteintech
Anti-RPRD1A Rabbit Polyclonal Antibody
Catalog # 10090-930
Supplier:  Proteintech
Supplier Number:  23652-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Regulation Of Nuclear Pre-MRNA Domain Containing 1A 
  • Antigen Symbol:
    RPRD1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    P15RS|P15INK4B-Related Protein
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10090-930
  • Supplier no.:
    23652-1-AP

Specifications

About this item

RPRD1A is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15(INK4b) and may have a role in cell cycle regulation. It contains a RAR domain that is involved in regulation of nuclear pre-mRNA, which suggests that P15RS may act as a nuclear regulation protein . It also interacts with phosphorylated C-terminal heptapeptide repeat domain (CTD) of the largest RNA polymerase II subunit POLR2A, and participates in dephosphorylation of the CTD. Besides, RPRD1A regulates cell cycle genes and attenuates WNT signaling , and acts as a negative regulator of cyclin-D1 (CCND1) and cyclin-E (CCNE1) in the cell cycle .

Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: RPRD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse