Order Entry
United States
ContactUsLinkComponent
Anti-WDR61 Rabbit Polyclonal Antibody
Anti-WDR61 Rabbit Polyclonal Antibody
Catalog # 10097-118
Supplier:  Proteintech
undefined
Anti-WDR61 Rabbit Polyclonal Antibody
Catalog # 10097-118
Supplier:  Proteintech
Supplier Number:  22536-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    WD Repeat Domain 61
  • Antigen Symbol:
    WDR61
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    WD repeat containing protein 61|WD repeat domain 61|Meiotic recombination REC14 protein homolog|SKI8|WDR61_HUMAN.|WDR 61|Recombination protein REC14|REC14
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10097-118
  • Supplier no.:
    22536-1-AP

Specifications

About this item

Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat