To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:PHD Finger Protein 20
- Antigen Symbol:PHF20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10092-192
- Supplier no.:22010-1-AP
Specifications
About this item
PHD finger protein 20 (PHF20),also known as NZF, TZP, GLEA2 or HCA58, is a 1,012 amino acid protein that localizes to the nucleus and contains one PHD-type zinc finger, one C2H2-type zinc finger and one A/T hook DNA-binding domain.PHF20 contains a tudor domain, which facilitates binding to di-methylated histone H4 Lys20. PHF20 is expressed in a variety of tissues, including liver, heart, lung, pancreas, spleen, testis and placenta, PHF20 is thought to function as a transcription factor, possibly playing a role in the development of cancers, including glioblastoma, lung cancer, colon cancer and ovarian cancer. PHF20 exists as three alternatively spliced isoforms (150kd,130kd and 120kd) which are encoded by a gene that maps to human chromosome 20 (22072714).
Western Blot: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: PHF20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse