Order Entry
United States
Orders LinkContactUsLinkComponent
Anti-GFRA2 Rabbit Polyclonal Antibody

Anti-GFRA2 Rabbit Polyclonal Antibody

150 µL
Catalog # 10081-802
Supplier:   Proteintech
Supplier #:  21973-1-AP-150UL
Anti-GFRA2 Rabbit Polyclonal Antibody
Catalog # 10081-802
Supplier:  Proteintech
Anti-GFRA2 Rabbit Polyclonal Antibody
Anti-GFRA2 Rabbit Polyclonal Antibody

150 µL

Price
$467.56
Log in for availability
Each (150µl)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

GFRA2, also named as GDNFRB, RETL2 and TRNR2, belongs to the GDNFR family. It is a receptor for neurturin. GFRA2 mediates the NRTN-induced autophosphorylation and activation of the RET receptor. It is a potent survival factor for central dopaminergic neurons, motor neurons, and several other populations of neurons in the central and peripheral nervous systems. The GDNF signal is mediated by a complex of the receptor tyrosine kinase RET and a glycosylphosphatidylinositol (GPI)-linked protein, GDNFRA. GFRA2 has three isoforms with MW 34-36 kDa, 39-41 kDa and 50-52 kDa. This antibody recognizes all the three isoforms.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: GFRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Specifications
  • Size:
    150 µL
  • Clonality:
    Polyclonal
  • Purification:
    Antigen affinity Purification
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cat. No.:
    10081-802
  • Reactivity:
    Human|Mouse
  • Western Blot:
    Yes
  • Shipping Temperature:
    Store at -20°C
  • Antibody Type:
    Primary
  • Antigen Symbol:
    GFRA2
  • Immunogen:
    Fusion Protein
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen Synonyms:
    GDNF family receptor alpha-2|NTNRA|RETL2|GDNFRB|GDNF receptor alpha-2|GDNF receptor beta|TRNR2|GFR alpha 2|NRTNR-ALPHA
  • Antigen Name:
    GDNF Family Receptor Alpha 2
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.