To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HBG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10088-076
- Supplier no.:19993-1-AP
Specifications
About this item
HBG1, also named as Hb F Agamma, HBGA, HBGR and HSGGL, belongs to the globin family. HBG1 makes up the fetal hemoglobin F, in combination with alpha chains. Some gamma variants can cause severe jaundice and cyanosis in premature and new born babies. The antibody is specific to HBG1 and HBG2. The antibody has no cross reaction to HBB and HBD.
Western Blot:recombinant proein, 1:1000-1:10000
Type: Primary
Antigen: HBG1-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human