To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Transmembrane Protein 176B
- Antigen Symbol:TMEM176B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-096
- Supplier no.:19825-1-AP
Specifications
About this item
TMEM176B (also known as LR8) is a transmembrane protein belongs to the MS4A family of proteins. It may play a role in the process of maturation of dendritic cells. TMEM176B was first discovered in human lung fibroblasts and is associated with human small cell lung carcinoma. Studies suggest that TMEM176B might be involved in cancer and might serve as targets for antiangiogenic therapy of cancer patients .
Western Blot:A549 Cells, 1:200-1:2000
Type: Primary
Antigen: TMEM176B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse