Order Entry
United States
ContactUsLinkComponent
 
Anti-POU3F3 Rabbit Polyclonal Antibody
  10092-608
undefined
Anti-POU3F3 Rabbit Polyclonal Antibody
  10092-608
 :  18999-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    POU Class 3 Homeobox 3
  • Antigen Symbol:
    POU3F3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    OTF-8|octamer-binding transcription factor 8|brain-specific homeobox/POU domain protein 1|oct-8|class 3|BRN1|octamer-binding protein 8|transcription factor 3|brain-1|brn-1|OTF8|POU domain
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10092-608
  • Supplier no.:
    18999-1-AP

 

 

OTF8, also named as POU3F3 and BRN1, belongs to the POU transcription factor family and Class-3 subfamily. The antibody is specific to OTF8.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: POU3F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat