To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NLR family member X1
- Antigen Symbol:NLRX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:MGC131937|MGC21025|OTTHUMP00000205769|NOD-like receptor X1|NOD9|NLRX1|nucleotide-binding oligomerization domain|FLJ21478|CLR11.3|NLR family|NLR family member X1|X1|NOD26|leucine rich repeat containing X1|DLNB26|NOD5|OTTHUMP00000205768
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10091-148
- Supplier no.:17215-1-AP
Specifications
About this item
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Heart Tissue, 1:200-1:2000
Type: Primary
Antigen: NLRX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat