Order Entry
United States
ContactUsLinkComponent
Anti-TCTN2 Rabbit Polyclonal Antibody
Anti-TCTN2 Rabbit Polyclonal Antibody
Catalog # 10095-750
Supplier:  Proteintech
undefined
Anti-TCTN2 Rabbit Polyclonal Antibody
Catalog # 10095-750
Supplier:  Proteintech
Supplier Number:  17053-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tectonic Family member 2
  • Antigen Symbol:
    TCTN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    MKS8|C12orf38|Tectonic-2|TCTN2|TECT2
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10095-750
  • Supplier no.:
    17053-1-AP

Specifications

About this item

TCTN2 (tectonic-2 or TECT2) is a type I membrane protein that belongs to the tectonic family, which consists of TCTN1, TCTN2 and TCTN3. Studies in mice suggest that tectonic may be involved in Hedgehog (Hh) signaling, and essential for ciliogenesis. Tectonic is component of the tectonic-like complex, a complex localized at the transition zone of primary cilia and acting as a barrier that prevents diffusion of transmembrane proteins between the cilia and plasma membranes. It exists two spilcing isoforms. The first aberrant transcript introduces 104 base pair from intron 13 and would delete 196 original amino acid, introduce two novel amino acids, and prematurely truncate the 697aa protein at residue number 504. The second transcript lacks exon 14 and would delete 195 original amino acids, introduce four novel amino acids, and prematurely truncate the 697 aa protein at residue number 507. The molecular weight of two isoforms are about 50kDa. We detected the 50kDa and 90kDa protein by immunoprecipitaion, but 50kDa protein may be the splicing isoform or non-specific band.

IP: RPE1 Cells(siRNA), 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:20-1:200

Type: Primary
Antigen: TCTN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat