Order Entry
United States
ContactUsLinkComponent
Anti-OCIAD1 Rabbit Polyclonal Antibody
Anti-OCIAD1 Rabbit Polyclonal Antibody
  10091-528
Anti-OCIAD1 Rabbit Polyclonal Antibody
  10091-528
 :  16634-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    OCIA Domain Containing 1
  • Antigen Symbol:
    OCIAD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    Ovarian carcinoma immunoreactive antigen|TPA018|OCIA domain-containing protein 1|OCIAD1|ASRIJ|OCIA
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10091-528
  • Supplier no.:
    16634-1-AP

 

 

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Liver, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: OCIAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat