To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dermokine
- Antigen Symbol:DMKN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:UNQ729/PRO1411|Dermokine|Epidermis-specific secreted protein SK30/SK89|UNQ729|ZD52F10|DMKN
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10085-572
- Supplier no.:16252-1-AP
Specifications
About this item
DMKN, also named as epidermis-specific secreted protein SK30/SK89, acts as a soluble regulator of keratinocyte differentiation. DMKN has 16 isoforms with MW 9-20 kDa, 35-38 kDa and 42-48 kDa.
Western Blot: Mouse ovary Tissue, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200
Type: Primary
Antigen: DMKN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat