To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Peroxisomal Biogenesis Factor 16
- Antigen Symbol:PEX16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10091-456
- Supplier no.:14816-1-AP
Specifications
About this item
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. Peroxin 16, also known as PEX16 or Peroxisomal biogenesis factor 16, is a 336 amino acid integral membrane protein that has a critical role in the biogenesis of peroxisomes. PEX16 together with PEX3 and PEX19 are specifically involved in peroxisomal membrane protein (PMP) import. Defects in the gene encoding Peroxin 16 are the cause of multiple peroxisome-related disorders, including Zellweger syndrome (ZWS), neonatal adrenoleukodystrophy (NALD), infantile Refsum disease (IRD), classical rhizomelic chondrodysplasia punctata (RCDP) and peroxisome biogenesis disorder complementation group 9 (PBD-CG9).
Western Blot: Human Liver Tissue, 1:500-1:5000
Type: Primary
Antigen: PEX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat