Anti-ISCU Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Iron-sulfur (Fe-S) clusters are required for the functions of mitochondrial aconitase, mammalian iron regulatory protein 1, and many other proteins in multiple subcellular compartments. Mitochondrial and cytosolic isoforms of the human Fe-S cluster scaffold protein, iron-sulfur cluster assembly enzyme (ISCU), is involved in activation of iron uptake, redistribution of intracellular iron, and decreased utilization of iron in Fe-S proteins.
Defects in ISCU gene are the cause of myopathy with exercise intolerance Swedish type (MEIS), which is characterized by lifelong severe exercise intolerance . Two ISCU isoforms ISCU1 (cytosolic, 15 kDa) and ISCU2 (mitochondrial, 18 kDa), were produced by alternative splicing, and ISCU2 is considered the canonical isoform1. ISCU2 protein is rapidly processed upon maturation . Proteintech’s ISCU antibody 14812-1-AP is raised against the full-length ISCU2 and is expected to detect 14 kDa,15 kDa and 18-19 kDa bands which has been reported in the previous observation .
Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:2000
Type: Primary
Antigen: ISCU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:150 µL
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Antigen affinity Purification
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- ImmunoFluorescence:Yes
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10088-986
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Store at -20°C
- Antibody Type:Primary
- Antigen Symbol:ISCU
- Immunogen:Recombinant Protein
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:NIFUN|NIFU|2310020H20Rik|hnifU|HML|ISU2
- Antigen Name:Iron-Sulfur Cluster Assembly Enzyme
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



