To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:High Mobility Group 20B
- Antigen Symbol:HMG20B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- ChIP:Yes
- Antigen Synonyms:BRAF25|SOXL|pp8857|SMARCE1r|PP7706|BRAF35|HMGXB2|HMGX2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10088-330
- Supplier no.:14582-1-AP
CoREST (corepressor of repressor element-1-silencing transcription factor) is a chromatin- modifying complex that was first identified as a regulator of neuronal gene expression, having a eseential role in neuronal differentiation . The CoREST complex is recruited by REST (RE1-silencing transcription factor) that recognizes the 21-nucleotide RE-1 sequence in the promoter of more than 1,700 different target genes in human and mouse . HMGB20B is one component of the CoREST complex, which mediates chromatin remodeling and repression of specific genes involved in neuronal differentiation through modulation of chromatin structure .
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Colon cancer, 1:20-1:200
Type: Primary
Antigen: HMG20B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat