To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:RNMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10093-826
- Supplier no.:13743-1-AP
Specifications
About this item
RNMT, also named as mRNA cap guanine-N7 methyltransferase or KIAA0398, is a 476 amino acid protein, which contains one mRNA cap 0 methyltransferase domain and belongs to the class I-like SAM-binding methyltransferase superfamily. mRNA cap 0 methyltransferase family. RNMT localizes in the nucleus and is widely expressed in various tissues. RNMT as a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5'-cap structure of mRNAs and binds RNA containing 5'-terminal GpppC.
Western Blot: Human Kidney Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200
Type: Primary
Antigen: RNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat