To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:A-kinase anchor protein 7 isoform gamma
- Antigen Symbol:AKAP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:AKAP18|AKAP15
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-718
- Supplier no.:12591-1-AP
Specifications
About this item
AKAP7, also named as AKAP15 and AKAP18, localize PKA in complexes with CaV1.2 and PLN, respectively. AKAP7 targets the cAMP-dependent protein kinase (PKA) to the plasma membrane, and permits functional coupling to the L-type calcium channel. AKAP7 has short isoform AKAP7 alpha and AKAP7 beta with MW 15-18 kDa; long isoform AKAP7 gamma with MW 37-42 kDa. AKAP7alpha was highly expressed only in brain and weakly in lung lysates from WT animals. This antibody can recognize all the isoforms of AKAP7.
IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; Western Blot: Human Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: AKAP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat