To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ring Finger Protein 7
- Antigen Symbol:RNF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:regulator of cullins 2|OTTHUMP00000180645|sensitive to apoptosis|SAG|ROC2|CKBBP1|zinc RING finger protein SAG
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10093-818
- Supplier no.:11905-1-AP
Specifications
About this item
SAG (sensitive to apoptosis gene protein ) is a novel zinc RING finger protein which is redox responsive and protects mammalian cells from apoptosis.It is shown to be localised in the cytoplasmand nucleus of the cell and is expressed in the heart, liver, skeletal muscle predominantly.SAG forms oligomers(55kd),dimer(28kd) and monomer(13kd) in the presence of different concentrations of the reductant.
Western Blot: Human Liver Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: RNF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat