Anti-GNAS Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide 1 (GNAS1) is the ubiquitously expressed heterotrimeric G protein that couples receptors to the effector enzyme adenylyl cyclase and is required for receptor-stimulated intracellular cAMP generation. Mutations of Gs(alpha) residues involved in the GTPase reaction that lead to constitutive activation are present in endocrine tumors, fibrous dysplasia of bone, and McCune-Albright syndrome. The molecular weight of Gs(alpha) protein is about 46 kDa.
Western Blot: Mouse Liver Tissue, 1:200-1:1000
Type: Primary
Antigen: GNAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:150 µL
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10087-686
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Store at -20°C
- Antibody Type:Primary
- Antigen Symbol:GNAS
- Immunogen:Recombinant Protein
- Conjugation:Unconjugated
- Antigen Synonyms:NESP|GSP|POH|GPSA|SCG6|GSA|C20orf45|AHO|GNAS1|SgVI
- Antigen Name:GNAS Complex locus
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



