Order Entry
United States
ContactUsLinkComponent
Bio-Clean Laboratory Detergent, Stanbio Laboratory
Bio-Clean Laboratory Detergent, Stanbio Laboratory
Catalog # 10152-744
Supplier:  Stanbio Laboratory
undefined
Bio-Clean Laboratory Detergent, Stanbio Laboratory
Catalog # 10152-744
Supplier:  Stanbio Laboratory
Supplier Number:  0602-380

Specifications

  • Description:
    Bio-Clean Laboratory Detergent
  • Environmentally Preferable:
  • Cat. No.:
    10152-744
  • Volume:
    3.8 L (1 gal.)
  • Supplier no.:
    0602-380

Specifications

About this item

Bio-Clean is a concentrate for cleaning laboratory instruments, glassware, surgical instruments, porcelain, metal, rubber and plastics

For normal cleaning, 5 mL dilutes to 1 gallon (3.785 L), thus each bottle will yield over 750 gallons (2839L) of cleaning power.