Technocut® Plus Disposable Safety Scalpels are top activated for ambidextrous use with a color-coded slider button for easy blade size identification
The fully extended blade enables clear line of sight and the fully enclosed front end enhances blade stability and security. Audible clicks help confirm the proper blade position with a final lock feature for safe handling and disposal.