Order Entry
United States
ContactUsLinkComponent
Bovine Serum Albumin (BSA)
Bovine Serum Albumin (BSA)
Catalog # 89508-826
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Bovine Serum Albumin (BSA)
Catalog # 89508-826
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Size:
    1 kg
  • Environmentally Preferable:
  • Storage Conditions:
    Refrigerator
  • Protease-free:
    Y
  • Protein Synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/Peptide Name:
    Bovine Serum Albumin
  • Color:
    light yellow
  • Cat. No.:
    89508-826
  • Supplier no.:
    AK1391-1000

Specifications

About this item

Bovine serum albumin, protease-free (protein stabilizing) may be used as a protein stabilizer for storage of working stocks of antibodies for flow cytometry, A BSA protein contains 583 amino acids, and has a molecular weight of 66.5 kDa.

  • Protease-free