Order Entry
United States
ContactUsLinkComponent
Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD16 + CD32 Rat Monoclonal Antibody [clone: 2.4G2]
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 10051-662
  • 10051-664
  • 10051-660
  • Antigen Symbol:
  • CD16 + CD32
  • CD16 + CD32
  • CD16 + CD32
  • Antigen Name:
  • FcR IgG Low Affinity 2
  • FcR IgG Low Affinity 2
  • FcR IgG Low Affinity 2
  • Antigen Synonyms:
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • CD32|Fc-gamma-RIIc|Fc gamma Receptor III|Fc-gamma RII-b|Fc gamma Receptor II|FcRII-c|FCGR3B|Fc-gamma-RIIb|FCGR3A|FCG2|Low affinity immunoglobulin gamma Fc region receptor II-c|CD32 receptor 2|Fc gamma Receptor 2|FCRIII|CD16A|Fc gamma Receptor 3|Fc-gamma RII-c|RP11-5K23.1|FCGR3|CDw32|FCRIIIA|FCR-10|FCGR2B|FCGR2|CD16B|FCGR2A|FCGRIII|FCGR2C|FcRII-b|Low affinity immunoglobulin gamma Fc region receptor II-b
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2.4G2
  • 2.4G2
  • 2.4G2
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 14130/14131
  • 14130/14131
  • 14130/14131
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
  • Immunofluorescence Microscopy
  • Immunofluorescence Microscopy
  • Immunofluorescence Microscopy

Specifications

Related Information

Frequently Bought Together