XGlove 8 Mil Extra-Thick Nitrile Glove, TechNiGlove
Supplier: Avantor
Certificates
About this item
TechNiGlove 9.5", 8 mil thick, powder-free nitrile gloves provide two times the puncture resistance of ordinary gloves.
- 8 mil thick
- Puncture-resistant
- Industrial use
- 1,000 gloves per case
TechNiGlove 8 mil, powder-free nitrile gloves provide hand and product protection in food, pharmaceutical, laboratory, and industrial applications. The XGlove resists a broad range of chemicals, offering protection from intermittent contact or splash protection when needed. The XGlove provides twice the puncture resistance of natural rubber disposable gloves of comparable thickness.
Ordering information: 1,000 gloves per case. Packaged 50 gloves per box, 20 boxes per case.
Frequently Bought Together