To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:regulator of G-protein signaling 12
- Antigen Symbol:RGS12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O14924
- Antigen Synonyms:RGS12
- Storage Buffer:Phosphate-Buffered Saline. No preservatives added.
- Molecular Weight:156.4 kDa (calculated)
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:1372-1447
- Purification:Antigen affinity-purified
- Cat. No.:10069-830
- Supplier no.:XW-7522
Specifications
About this item
Regulator of G protein signalling 12 , RGS12, domain aa 1372-1447. This protein negatively regulates G protein-coupled receptor signalling.
RGS12 (C-76) antibody can be used for the detection of RGS12 (C-76) by Western blot, may also work for IHC and ICC.
Antigen affinity-purified
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RGS12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat