To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:fatty acid binding protein 4, adipocyte
- Antigen Symbol:FABP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mL
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P15090
- Antigen Synonyms:FABP4
- Storage Temperature:2-8 °C for one week and –20 °C as an undiluted liquid for up to two years.
- Shipping Temperature:4 °C
- Immunogen:Human ALBP peptide.
- Cat. No.:10068-070
- Supplier no.:XG-6174
Specifications
About this item
This antibody can be used at 1:200-1:5000 dilutions for Western Blot analysis and for other immuno-assays. Customers need to optimize the dilution when using the Abs in IHC or ICC.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human