Order Entry
United States
ContactUsLinkComponent
Anti-EIF4E Rabbit Polyclonal Antibody
Anti-EIF4E Rabbit Polyclonal Antibody
Catalog # 10071-904
Supplier:  Prosci
undefined
Anti-EIF4E Rabbit Polyclonal Antibody
Catalog # 10071-904
Supplier:  Prosci
Supplier Number:  79-374
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    eukaryotic translation initiation factor 4E
  • Antigen Symbol:
    EIF4E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    P06730
  • Antigen Synonyms:
    EIF4E
  • Storage Buffer:
    Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
  • Molecular Weight:
    25 kDa
  • Storage Temperature:
    Store antibody at –20 °C for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    eIF4E (Ab-209) antibody was raised against a peptide sequence around aa. 207~211 (S-G-S-T-T) derived from Human eIF4E.
  • Purification:
    Antibodies were purified by affinity-chromatography using epitope-specific peptide.
  • Cat. No.:
    10071-904
  • Supplier no.:
    79-374

Specifications

About this item

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.

Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat