Order Entry
United States
ContactUsLinkComponent
Anti-Heartland Virus Rabbit Polyclonal Antibody
Anti-Heartland Virus Rabbit Polyclonal Antibody
Catalog # 10061-936
Supplier:  Prosci
undefined
Anti-Heartland Virus Rabbit Polyclonal Antibody
Catalog # 10061-936
Supplier:  Prosci
Supplier Number:  7435
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Glycoprotein polyprotein
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    J3WAX0
  • Storage Buffer:
    Heartland virus glycoprotein 2 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular Weight:
    Predicted: 55 kDa
  • Storage Temperature:
    Heartland virus glycoprotein 2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Heartland virus glycoprotein 2 antibody was raised against a 19 amino acid peptide near the center of the Heartland virus glycoprotein 2. The immunogen is located within amino acids 860 - 910 of Heartland Virus Glycoprotein 2.
  • Purification:
    Heartland Virus Glycoprotein 2 Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    10061-936
  • Supplier no.:
    7435

Specifications

About this item

Heartland Virus Glycoprotein 2 Antibody: The Heartland virus is a newly identified member of the phlebovirus genus of the Bunyaviridae family. Initial reports indicate that symptoms may include fever, fatigue, diarrhea, thrombocytopenia, and leukopenia. The Bunyaviridae family of viruses are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The glycoprotein RNA encodes a polyprotein that is cleaved into the G1 and G2 proteins.

Heartland virus glycoprotein 2 antibody can detect 10 ng Heartland virus glycoprotein 2 in ELISA at 1 ?g/mL.

Heartland Virus Glycoprotein 2 Antibody is affinity chromatography purified via peptide column.

Heartland virus glycoprotein 2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Glycoprotein polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus