To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:cholecystokinin A receptor
- Antigen Symbol:CCKAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Monkey,Gibbon,Gorilla,Human,Dog
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:P32238
- Antigen Synonyms:CCKAR
- Storage Buffer:PBS, 0.1% sodium azide.
- Storage Temperature:Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Synthetic 16 amino acid peptide from human CCKAR.
- Tested Applications:Immunoaffinity Chromatography
- Purification:Immunoaffinity Chromatography
- Cat. No.:10080-764
- Supplier no.:71-192
Specifications
About this item
CCKAR Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 10 ?g/mL.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CCKAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Human, Monkey