Order Entry
United States
ContactUsLinkComponent
Anti-ST14 Rabbit Polyclonal Antibody
Anti-ST14 Rabbit Polyclonal Antibody
  10080-306
 :  Prosci
undefined
Anti-ST14 Rabbit Polyclonal Antibody
  10080-306
 :  Prosci
 :  70-824
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    suppression of tumorigenicity 14 (colon carcinoma)
  • Antigen Symbol:
    ST14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mL
  • Environmentally Preferable:
  • Gene ID:
    Q9Y5Y6
  • Antigen Synonyms:
    ST14
  • Storage Buffer:
    PBS (without Mg2+, Ca2+), pH 7.4, 150 mM sodium chloride, 0.02% sodium azide, 50% glycerol
  • Storage Temperature:
    Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Synthetic peptide from human ST14.
  • Tested Applications:
    Immunoaffinity Chromatography
  • Purification:
    Immunoaffinity Chromatography
  • Cat. No.:
    10080-306
  • Supplier no.:
    70-824

 

 

ST14 / MatrImmunoprecipitationtase Antibody can be used for ELISA starting at 1:10000, Immunofluorescence starting at 1:100 - 1:500, Immunohistochemistry in paraffin embeded tissues starting at 10?g/mL, and Western Blotting starting at 1:500 - 1:1000.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ST14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human