To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:growth hormone 1
- Antigen Symbol:GH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Bovine
- Size:0.05 mL
- Environmentally Preferable:
- Gene ID:P01241
- Antigen Synonyms:GH1
- Storage Temperature:Short term: 4 °C. Long term: Store at –20 °C. Avoid freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Bovine growth hormone.
- Purification:Antiserum
- Cat. No.:10080-082
- Supplier no.:70-652
Specifications
About this item
GH1 / Growth Hormone Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 1:100.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human