Order Entry
United States
ContactUsLinkComponent
Anti-TGM2 Rabbit Polyclonal Antibody
Anti-TGM2 Rabbit Polyclonal Antibody
Catalog # 10074-552
Supplier:  Avantor
undefined
Anti-TGM2 Rabbit Polyclonal Antibody
Catalog # 10074-552
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    transglutaminase 2
  • Antigen Symbol:
    TGM2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.05 mL
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    P21980
  • Antigen Synonyms:
    TGM2
  • Storage Buffer:
    PBS, pH 7.4, 0.2% BSA, 0.1% sodium azide.
  • Storage Temperature:
    Store Transglutaminase 2 antibody at 4 °C or –20 °C. As with all antibodies avoid freeze/thaw cycles.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Transglutaminase 2 antibody was raised against recombinant Transglutaminase 2 (Human).
  • Purification:
    Protein A Column
  • Cat. No.:
    10074-552
  • Supplier no.:
    49-796

Specifications

About this item

Transglutaminases (TGases) are calcium-dependent enzymes catalysing the post-translational cross-linking of proteins.

Transglutaminase 2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Protein A Column

PBS, pH 7.4, 0.2% BSA, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TGM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat