Order Entry
United States
ContactUsLinkComponent
Anti-CCNL1 Rabbit Polyclonal Antibody
Anti-CCNL1 Rabbit Polyclonal Antibody
Catalog # 10079-154
Supplier:  Avantor
undefined
Anti-CCNL1 Rabbit Polyclonal Antibody
Catalog # 10079-154
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    cyclin L1
  • Antigen Symbol:
    CCNL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Gene ID:
    Q9UK58
  • Antigen Synonyms:
    CCNL1
  • Storage Temperature:
    Store Cyclin L1 antibody at 4 °C or –20 °C. As with all antibodies avoid freeze/thaw cycles.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    RT
  • Immunogen:
    Cyclin L1 antibody was raised against amino acids 579 - 593 of Cyclin L1 (Human).
  • Purification:
    Protein A Column
  • Cat. No.:
    10079-154
  • Supplier no.:
    49-663

Specifications

About this item

Cyclin L (also referred to as CCNL) is encoded by two highly related genes Cyclin L1 and Cyclin L2 (CCNL1 and CCNL2, respectively). Cyclin L has been shown to associate with the PITSLRE kinase and is involved in pre-mRNA processing. Both Cyclin L gene products can be alternatively spliced to produce two isoforms (known as 1 and 2 or alpha and beta). Cyclin L2 is ubiquitous, expressed at much higher levels than Cyclin L1, and thus is likely the major partner for the CDK11p110 protein kinase.

Cyclin L1 antibody can be used in Western Blot, and immunohistochemistry starting at 1:100 - 1:200.

Protein A Column

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat