To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:calmodulin 1 (phosphorylase kinase, delta)
- Antigen Symbol:CALM1
- Clonality:Monoclonal
- Clone:[J4D8]
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P62158
- Antigen Synonyms:CALM1
- Storage Buffer:PBS, pH 7.4, 0.1% sodium azide
- Storage Temperature:Short term: 4 °C. Long term: Store at –20 °C. Avoid freeze-thaw cycles.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human Calmodulin (1-149aa) purified from E. coli.
- Purification:Protein G Column
- Cat. No.:10077-738
- Supplier no.:48-075
Specifications
About this item
Calmodulin antibody can be used in immunohistochemistry starting at 5 ?g/mL.
PBS, pH 7.4, 0.1% sodium azide
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CALM1
Clonality: Monoclonal
Clone: [J4D8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human