To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:nuclear receptor subfamily 0, group B, member 1
- Antigen Symbol:NR0B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P51843
- Antigen Synonyms:NR0B1
- Storage Buffer:DAX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:DAX1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of DAX1.
- Purification:DAX1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10115-866
- Supplier no.:45-461
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50-55 kDa band observed in human ovary and human testis lysates (calculated MW of 51.7 kDa according to NP_000466.2). Recommended concentration: 0.5-1.5 ?g/mL.
DAX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: NR0B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human